Learn More
N-Boc-D-proline, 98+%, Thermo Scientific Chemicals Product Code.: 11342736
N-Boc-D-proline, 98+%, Thermo Scientific Chemicals
Product Code. 11342736
5 g
Click to view available options
Quantity:
1 g
5 g
Product Code. 11342736
Brand: Thermo Scientific Alfa Aesar L08826.06
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 37784-17-1
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.249
MDL Number MFCD00063226
InChI Key ZQEBQGAAWMOMAI-SSDOTTSWSA-N
Synonym n-boc-d-proline, boc-d-proline, boc-d-pro-oh, r-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butoxycarbonyl-d-proline, 2r-1-tert-butoxy carbonyl pyrrolidine-2-carboxylic acid, n-t-boc-d-proline, boc-d-pro, n-alpha-t-boc-d-proline, 1-tert-butoxycarbonyl-d-prolineShow MoreShow Less
PubChem CID 688022
IUPAC Name (2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCC1C(=O)O
Specifications
Melting Point 133°C to 135°C
Quantity 5 g
Beilstein 479316
Optical Rotation +61° (c=1 in Acetic Acid)
Formula Weight 215.25
Percent Purity ≥98%
Chemical Name or Material N-Boc-D-proline
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10197181 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberL08826.06,L08826.03
Lot NumberB04X001 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL08826.06,L08826.03
Lot Number10148926 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL08826.06,L08826.03
Lot NumberE18U022 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL08826.06,L08826.03
Lot NumberG18T015 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL08826.06,L08826.03
RUO – Research Use Only
Product Content Correction
Product Title
N-Boc-D-proline, 98+%, Thermo Scientific Chemicals >