missing translation for 'onlineSavingsMsg'
Learn More
N-Boc-L-histidine methyl ester, 95%, Thermo Scientific Chemicals Product Code.: 15409068

N-Boc-L-histidine methyl ester, 95%, Thermo Scientific Chemicals

Product Code. 15409068
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15409068

Brand: Thermo Scientific Alfa Aesar H62545.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2488-14-4
Molecular Formula C12H19N3O4
Molecular Weight (g/mol) 269.301
MDL Number MFCD00167635
InChI Key RWEVEEORUORBAX-VIFPVBQESA-N
Synonym boc-his-ome, nalpha-boc-l-histidine methyl ester, l-histidine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propanoate, n-boc-l-histidine methyl ester, na-boc-l-histidine methyl ester, n-tert-butoxycarbonyl-l-histidine methyl ester, l-histidine,n-1,1-dimethylethoxy carbonyl-, methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propanoate, s-methyl 2-tert-butoxycarbonyl amino-3-1h-imidazol-5-yl propanoateShow MoreShow Less
PubChem CID 7017200
IUPAC Name methyl (2S)-3-(1H-imidazol-5-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(CC1=CN=CN1)C(=O)OC

Specifications

Quantity 5 g
Formula Weight 269.3
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-L-histidine methyl ester

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ30K025 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog NumberH62545.03,H62545.06
Lot NumberU08D035 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62545.03,H62545.06,H62545.MD
Lot NumberR13H017 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62545.03,H62545.06,H62545.MD
Lot NumberY02G016 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62545.03,H62545.06,H62545.MD
Lot NumberR21E047 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62545.03,H62545.06,H62545.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Boc-L-histidine methyl ester, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.