missing translation for 'onlineSavingsMsg'
Learn More
N-Boc-trans-4-hydroxy-L-proline methyl ester, 97% Product Code.: 15485198

N-Boc-trans-4-hydroxy-L-proline methyl ester, 97%

Product Code. 15485198
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15485198

Brand: Thermo Scientific Alfa Aesar H59027.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 74844-91-0 | C11H19NO5 | 245.28 g/mol

It is used in the synthesis of For-Met-Leu-Phe-OMe (fMLF-OMe) analogues based on Met residue replacement by 4-amino-proline scaffold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of For-Met-Leu-Phe-OMe (fMLF-OMe) analogues based on Met residue replacement by 4-amino-proline scaffold.

Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74844-91-0
Molecular Formula C11H19NO5
Molecular Weight (g/mol) 245.28
MDL Number MFCD00076981
InChI Key MZMNEDXVUJLQAF-SFYZADRCSA-N
Synonym boc-hyp-ome, n-boc-trans-4-hydroxy-l-proline methyl ester, 2s,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, boc-l-hydroxyproline methyl ester, boc-hydroxyproline-ome, methyl trans-1-boc-4-hydroxy-l-prolinate, n-tert-butoxycarbonyl-trans-4-hydroxy-l-proline methyl ester, trans-1-tert-butoxycarbonyl-4-hydroxy-l-proline methyl ester, n-boc-trans-4-hydroxyl-l-proline methyl esterShow MoreShow Less
PubChem CID 2734883
SMILES COC(=O)[C@@H]1C[C@@H](O)CN1C(=O)OC(C)(C)C

Specifications

Melting Point 92°C to 96°C
Quantity 1 g
Solubility Information Soluble in chloroform,dichloromethane and ethyl acetate.
Optical Rotation −65° (c=1 in Chloroform)
IUPAC Name 1-tert-butyl 2-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
Formula Weight 245.28
Percent Purity 97%
Chemical Name or Material N-Boc-trans-4-hydroxy-L-proline methyl ester

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Product Code
Lot Number81200984 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH59027.06,H59027.03
Lot NumberR23C030 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH59027.06,H59027.03
Lot NumberY22D011 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH59027.06,H59027.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Boc-trans-4-hydroxy-L-proline methyl ester, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.