Learn More
N-Fmoc-L-glutamic acid 5-benzyl ester, 95%, Thermo Scientific™
Product Code. 15489258
Click to view available options
Quantity:
5 g
25 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 123639-61-2
Molecular Formula C27H25NO6
Molecular Weight (g/mol) 459.498
MDL Number MFCD00065642
InChI Key HJJURMMMGPQIQP-DEOSSOPVSA-N
Synonym fmoc-glu obzl-oh, fmoc-l-glutamic acid-gamma-benzyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid, fmoc-l-glutamic acid 5-benzyl ester, 2s-5-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, pubchem10013, fmoc-glu obzl-oh hplc, n-9-fluorenylmethoxycarbonyl-l-glutamic acid-gamma-benzyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-benzyl ester, 2s-2-fluoren-9-ylmethoxy carbonylamino-4-benzyloxycarbonyl butanoic acid
PubChem CID 13966932
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Specifications
Melting Point (decomposition)
Quantity 5 g
Beilstein 3576722
Optical Rotation +12° (c=1 in Chloroform)
Formula Weight 459.49
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-glutamic acid 5-benzyl ester