missing translation for 'onlineSavingsMsg'
Learn More
N-Methyl-N-(4-pyridin-4-ylbenzyl)amine, 90%, Thermo Scientific™ Product Code.: 10526153

N-Methyl-N-(4-pyridin-4-ylbenzyl)amine, 90%, Thermo Scientific™

Product Code. 10526153
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 10526153

Brand: Thermo Scientific Maybridge CC35446DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 852180-64-4
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.269
InChI Key MDKJAMMSIZLRBH-UHFFFAOYSA-N
Synonym n-methyl-n-4-pyridin-4-ylbenzyl amine, n-methyl-4-pyridin-4-yl benzylamine, methyl 4-pyridin-4-yl phenyl methyl amine, n-methyl-1-4-pyridin-4-yl phenyl methanamine, n-methyl-1-4-pyridin-4-ylphenyl methanamine, methyl 4-4-pyridyl phenyl methyl amine, benzenemethanamine,n-methyl-4-4-pyridinylShow MoreShow Less
PubChem CID 7060572
IUPAC Name N-methyl-1-(4-pyridin-4-ylphenyl)methanamine
SMILES CNCC1=CC=C(C=C1)C2=CC=NC=C2

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 198.27
Percent Purity 90%
Chemical Name or Material N-Methyl-N-(4-pyridin-4-ylbenzyl)amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Methyl-N-(4-pyridin-4-ylbenzyl)amine, 90%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.