Learn More
N,N-Dimethyl-n-octylamine, 95%, Thermo Scientific Chemicals
Description
N,N-Dimethyl-n-octylamine is used as ion-pairing agent in the quantitative estimation of thiazinamium methylsulphate in a pharmaceutical preparation by HPLC. It acts as a counterion in mobile phase for separation of tetracycline, tetracycline analogs and their potential impurities by reversed-phase ion-pair chromatography. Further, it is used in the study of chiral separations by complexation with proteins in capillary zone electrophoresis. In addition to this, it is used as a mobile phase in direct resolution and quantitation of (R)- and (S)-disopyramide
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N,N-Dimethyl-n-octylamine is used as ion-pairing agent in the quantitative estimation of thiazinamium methylsulphate in a pharmaceutical preparation by HPLC. It acts as a counterion in mobile phase for separation of tetracycline, tetracycline analogs and their potential impurities by reversed-phase ion-pair chromatography. Further, it is used in the study of chiral separations by complexation with proteins in capillary zone electrophoresis. In addition to this, it is used as a mobile phase in direct resolution and quantitation of (R)- and (S)-disopyramide
Solubility
Slightly miscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acids and copper.

Specifications
Specifications
Melting Point | -57°C |
Density | 0.765 |
Boiling Point | 195°C |
Flash Point | 65°C (149°F) |
Refractive Index | 1.4243 |
Quantity | 50 g |
UN Number | UN2922 |
Beilstein | 1698081 |
Sensitivity | Air Sensitive |
Solubility Information | Slightly miscible with water. |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0456675 | Certificate of Analysis | 30/08/2024 | A12546.18,A12546.09,A12546.30 |
A0455550 | Certificate of Analysis | 19/10/2023 | A12546.18,A12546.09,A12546.30 |
A0434757 | Certificate of Analysis | 19/10/2022 | A12546.18,A12546.09,A12546.30 |
10130736 | Certificate of Analysis | 22/11/2021 | A12546.18,A12546.09,A12546.30 |
E19Q25 | Certificate of Analysis | 22/11/2021 | A12546.18,A12546.09,A12546.30 |
Safety and Handling



- N,N-Dimethyl-n-octylamine
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : N,N-Dimethyl-n-octylamine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.