Learn More
Memmert™ N2 Pressure Reducing Valve to DIN EN ISO 2503
N2 pressure reducing valve with gas cylinder monitor
Brand:  Memmert™ ICO150E06162
 View more versions of this product
Product Code. 17129964
  • 2570.00 DKK / Each
Estimated Shipment: 26-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Requires option T6
  • Includes gas cylinder monitor
Specifications
Specifications
ICO150
N2 Pressure Reducing Valve
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ N2 Pressure Reducing Valve to DIN EN ISO 2503 > ICO150