Learn More
N4-Acetylcytidine triphosphate sodium, MedChemExpress
MedChemExpress N4-Acetylcytidine triphosphate sodium is efficiently used as a substrate in T7 Polymerase-catalyzed in vitro transcription and it can be incorporated into multiple templates.
Brand: Medchem Express HY-111815A
This item is not returnable.
View return policy
Description
N4-Acetylcytidine triphosphate sodium is efficiently used as a substrate in T7 Polymerase-catalyzed in vitroSpecifications
N4-Acetylcytidine triphosphate sodium | |
Research | |
5 mg | |
H2O : ≥ 250 mg/mL (407.75 mM) | |
613.12 | |
Research |
Off-White | |
C11H14N3Na4O15P3 | |
ac4CTP sodium | |
O[C@@H]([C@H]([C@H](N1C(N=C(C=C1)NC(C)=O)=O)O2)O)[C@H]2COP(OP(OP(O[Na])(O[Na])=O)(O[Na])=O)(O[Na])=O | |
613.12 | |
Solid |
Research purposes only