Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth Opaque Amber HDPE Bottles with Closure: Shrink-Wrapped Trays

Experience easy handling with Nalgene Opaque Amber HDPE Bottles, conveniently packaged in shrink-wrap trays ready to feed into filling lines for various reagents.
Brand: Thermo Scientific™ 322085-0001
7214.75 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Experience easy handling with Thermo Scientific™ Nalgene Opaque Amber HDPE Bottles, conveniently packaged in shrink-wrap trays ready to feed into filling lines for various reagents. Bottles include polypropylene closures.
- Opaque amber container provides protection for light-sensitive reagents
- Bottles are packed in rigid trays, allowing for easy handling
- Opaque amber HDPE with opaque amber polypropylene closures
- Leakproofâ€
Ordering Information:
Bottles are packed in shrink-wrapped trays with closures packed in separate polybags within the same carton. †The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
1 oz., 30 mL | |
1 oz. | |
Narrow-Mouth Packaging Bottles | |
HDPE | |
70 | |
20 mm | |
Yes | |
Narrow | |
61 mm | |
Polypropylene | |
34 mm | |
No |
30 mL | |
Opaque Amber | |
14 mm | |
1050 | |
Opaque Amber | |
Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition | |
No | |
Round | |
Narrow Mouth Bottle | |
20-415 mm | |
Screw (Bulk Packed, Unassembled) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.