Learn More
Thermo Scientific™ Nalgene™ PVDF True Union Clamps
Repeatedly and reliably seal carboys with Thermo Scientific™ Nalgene™ PVDF True Union Clamps with silicone gaskets and end caps.
Brand:  Thermo Scientific™ 2670-0150
581.95 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10506171
  • 600.00 DKK / Each
Only null left
Estimated Shipment: 07-10-2025
Description
Repeatedly and reliably seal carboys with Thermo Scientific™ Nalgene™ PVDF True Union Clamps with silicone gaskets and end caps. Molded of PVDF fluoride, these threaded clamps are a fully autoclavable clamp system.
  • Available in three sizes and with Mini and Tri-Clover clamp types
  • Threaded and autoclavable
  • Recommended for use with Nalgene™ Sanitary Carboys (Cat. Nos. 2261, 2630 and 2640) and Nalgene™ End Caps (Cat. Nos. 2688 and 2689)
  • Recommended for use with Nalgene™ Autoclavable Carboys with Spigot (Cat. No. 2319)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1/Pk., 1/Cs.
PVDF
1.5 in.
Tri Clover
Nalgene™ Sanitary Carboys, Nalgene™ Autoclavable Carboys with Spigot
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ PVDF True Union Clamps > 1.5 in. (38mm)
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.