Learn More
Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom: Sterile
Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing and storage.
651.00 DKK - 2430.00 DKK
Specifications
Material | PETG |
---|---|
No. per Pack | 1 |
Shape | Erlenmeyer |
Surface Coating | None |
Autoclavable | Not Autoclavable |
Product Code | Brand | Capacity (Metric) | Closure Size | Closure Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Closure Type | Price | Quantity & Availability | ||||
10266432
|
Thermo Scientific™
4115-0125 |
125 mL | 38-430 mm | Vented |
2430.00 DKK
Pack of 24 |
Estimated Shipment: 07-07-2025
Log in to see stock. |
||||
10592033
|
Thermo Scientific™
4115-0250 |
250 mL | 38-430 mm | Vented |
1335.00 DKK
Pack of 12 |
In Stock
Log in to see stock. ![]() |
||||
10686673
|
Thermo Scientific™
4115-0500 |
500 mL | 45-430 mm | Vented |
1590.00 DKK
Pack of 12 |
In Stock
Log in to see stock. ![]() |
||||
10357302
|
Thermo Scientific™
4115-1000 |
1000 mL | 45-430 mm | Vented |
892.00 DKK
Pack of 6 |
In Stock
Log in to see stock. ![]() |
||||
10473582
|
Thermo Scientific™
4115-2000 |
2000 mL | 45-430 mm | Vented |
872.00 DKK
Pack of 4 |
In Stock
Log in to see stock. |
||||
10156192
|
Thermo Scientific™
4115-2800 |
2800 mL | 70 mm | Vented |
1341.60 DKK
Pack of 4 |
In Stock
Log in to see stock. ![]() |
||||
10465622
|
Thermo Scientific™
4112-0125 |
125 mL | 38-430 mm | Solid, non-vented |
1710.00 DKK
Pack of 24 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
||||
10292032
|
Thermo Scientific™
4112-0250 |
250 mL | 38-430 mm | Solid, non-vented |
783.40 DKK
Pack of 12 |
In Stock
Log in to see stock. |
||||
10179661
|
Thermo Scientific™
4112-0500 |
500 mL | 45-430 mm | Solid, non-vented |
1160.00 DKK
Pack of 12 |
In Stock
Log in to see stock. |
||||
10475622
|
Thermo Scientific™
4112-1000 |
1000 mL | 45-430 mm | Solid, non-vented |
662.00 DKK
Pack of 6 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||
10385012
|
Thermo Scientific™
4112-2000 |
2000 mL | 45-430 mm | Solid, non-vented |
651.00 DKK
Pack of 4 |
In Stock
Log in to see stock. |
||||
10228732
|
Thermo Scientific™
4112-2800 |
2800 mL | 70 mm | Solid, non-vented |
1215.00 DKK
Pack of 4 |
In Stock
Log in to see stock. |
||||
Description
Save preparation time and avoid contamination risk with Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
Features:- Sterile and single use: Reduce contamination risk
- BPA-free
- Collapses when autoclaved: Reduce bio hazardous waste
- Graduated for quick volume assessment
- Regulatory documentation available on request
Specifications
PETG | |
Erlenmeyer | |
Not Autoclavable | |
Yes | |
Plain |
1 | |
None | |
Single-use | |
Sterile |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.