Learn More
Thermo Scientific™ Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure: Bulk Pack

Ship and store diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals in these light-weight bottles which are leakproof when used with Nalgene closures.
Brand: Thermo Scientific™ 312187-0004
4789.94 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Ship and store diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals with Thermo Scientific™ Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure. These durable, autoclavable bottles are designed for shipping and storage.
- Wide mouth allows for easy filling and dispensing of powders
- Injection blow molded for uniform wall thickness and strength
- Made from high-purity, food-grade resins; no plasticizers, fillers or extenders
- Polypropylene closures included
- Leakproofâ€
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton. †The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
4 oz., 125 mL | |
4 oz. | |
Wide-Mouth Packaging Bottles | |
PPCO | |
500 | |
38 mm | |
Yes | |
No | |
Round | |
Packer Bottle | |
3.92 in. (With Closure); 3.78 in. (Without Closure) | |
38-415 mm | |
Screw (Bulk Packed, Unassembled) |
125 mL | |
Translucent | |
28 mm | |
500 | |
Natural, translucent | |
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
Autoclavable | |
Wide | |
99 mm | |
1.99 in. | |
Polypropylene | |
50 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
†The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.