Learn More
Gibco™ NaOH, 1.0 M Product Code.: 17810935
Gibco™ NaOH, 1.0 M
Product Code. 17810935
200 L, 1.0 M, 200 L
Click to view available options
Quantity:
20 L
200 L
Concentration:
0.1 M
0.5 M
1.0 M
Unit Size:
20 L
200 L
Product Code. 17810935
Brand: Gibco™ A4782902
Gibco NaOH solutions offer more options for supply security and dual sourcing of the highest volume process liquid in bioprocessing manufacturing.
Gibco NaOH solutions offer more options for supply security and dual sourcing of the highest volume process liquid in bioprocessing manufacturing. Available in three different grades and two off-the-shelf sizes, these solutions provide outsourcing efficiencies to help reclaim time, space, and other core resources.
Features of Gibco NaOH solutions include:
  • 0.1 M, 0.5 M, and 1 M concentrations
  • Multi-compendial grade raw materials
  • cGMP manufacturing to 21CFR820
  • 20 L and 200 L ready-to-use single-use bioprocessing containers
Flexibility
Gibco NaOH solutions provide outsourcing efficiencies for customers manufacturing internally but seeking to partially outsource, as well as for customers with batch sizes that do not justify custom ordering and customers already outsourcing to Thermo Fisher Scientific but seeking additional flexibility. They are produced in the most commonly used concentrations and single-use containers.
Applications of Gibco NaOH solutions
  • pH adjustment
  • Facility/equipment sanitization
  • Chromatography column cleaning, sanitization and storage
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2
Melting Point 0°C
Boiling Point 100°C
Color Clear
Concentration 1.0 M
pH ≥12 to ≤14
Quantity 200 L
Molecular Formula HNaO
Packaging Bioprocess Container
Synonym Caustic soda, Acidity regulatory, Lye or Soda lye
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES [OH-].[Na+]
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40.00
Percent Purity 95 to 100%
Grade Compendial
Show More Show Less
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Product Content Correction
Product Title
Gibco™ NaOH, 1.0 M >
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado