Learn More
BRAND™ Narrow Neck Erlenmeyer Flask
Narrow Neck Erlenmeyer Flask features a beaded rim and graduation marks.
Brand: BRAND™ 92724
701.33 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Borosilicate Glass 3.3
- DIN ISO 1773
- With beaded rim and graduation
Specifications
100 mL | |
Narrow | |
Beaded |
Borosilicate Glass (3.3) | |
Yes |
Special Offers
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now