Learn More
Naspm trihydrochloride, MedChemExpress
MedChemExpress Naspm trihydrochloride (1-Naphthylacetyl spermine trihydrochloride), a synthetic analogue of Joro spider toxin, is a calcium permeable AMPA (CP-AMPA) receptors antagonist.

1107.84 DKK - 9265.54 DKK
Specifications
Chemical Name or Material | Naspm trihydrochloride |
---|---|
Physical Form | Solid |
CAS | 1049731-36-3 |
Color | Pale Yellow |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16461437
![]() |
Medchem Express
HY-12506A |
50 mg |
5267.25 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16415228
![]() |
Medchem Express
HY-12506A |
10 mg |
1490.55 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16407947
![]() |
Medchem Express
HY-12506A |
5 mg |
1107.84 DKK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16405906
![]() |
Medchem Express
HY-12506A |
100 mg |
9265.54 DKK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Naspm trihydrochloride (1-Naphthylacetyl spermine trihydrochloride), a synthetic analogue of Joro spider toxin, is a calcium permeable AMPA (CP-AMPA) receptors antagonist.Specifications
Naspm trihydrochloride | |
1049731-36-3 | |
Research | |
Neuroscience-Neuromodulation | |
H2O : 50 mg/mL (104.19 mM; Need ultrasonic) ∣DMSO : 6.4 mg/mL (13.34 mM; Need ultrasonic) | |
479.91 | |
98.0% |
Solid | |
Pale Yellow | |
C22H37Cl3N4O | |
1-Naphthylacetyl spermine trihydrochloride | |
[H]Cl.[H]Cl.[H]Cl.O=C(NCCCNCCCCNCCCN)CC1=C2C=CC=CC2=CC=C1 | |
479.91 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.