Learn More
Thermo Scientific™ National Screw Thread Caps with Bonded Septa
Effectively eliminate septa push-through with Thermo Scientific™ National Screw Thread Caps with Bonded Septa.
Brand:  Thermo Scientific™ C4013-65A
314.52 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10006704
  • 317.00 DKK / Pack of 100
Only null left
In Stock
Description
Effectively eliminate septa push-through with Thermo Scientific™ National Screw Thread Caps with Bonded Septa.
  • Septa and polypropylene caps bonded together for an integral system
  • Eliminates septa push-through
  • Available with choice of GPI thread sizes and preslit septa for easier thin-gauge needle penetration
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black; Bonded Red 0.005 in. PTFE/White Silicone Septa; GPI Thread Finish: 8-425
PTFE, Silicone, Polypropylene
Bonded Red PTFE/White Silicone, 0.005 in.
8-425
Black
No
National
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ National Screw Thread Caps with Bonded Septa > Black; Bonded Red 0.005 in. PTFE/White Silicone Septa; GPI Thread Finish: 8-425