Learn More
Thermo Scientific™ National Vial Crushers
Reduce waste disposal costs by reducing the total volume of waste with the Thermo Scientific™ National Vial Crusher.
Brand:  Thermo Scientific™ C4040-VC
100266.68 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10520136
  • 100770.00 DKK / Each
Only null left
Estimated Shipment: 29-09-2025
Description
Reduce waste disposal costs by reducing the total volume of waste with the Thermo Scientific™ National Vial Crusher.
  • Breaks and separates liquid from all 1mL to 100mL vials and ampules
  • Capable of processing approximately 5000 2mL vials per hour
  • Safe use in a fume hood or connect to a vacuum for limited exposure to flammable or toxic vapors
  • Environmentally responsible waste handling—crushed glass is automatically separated from solvent for separate disposal
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Manual Benchtop Vial Crusher, 241/2H x 81/2W x 231/2 in. D
National
Manual Benchtop Vial Crusher
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ National Vial Crushers > Manual benchtop vial crusher