missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1706.22 DKK - 3498.83 DKK
Specifications
Antigen | NDUFS3 |
---|---|
Applications | Western Blot, Immunohistochemistry, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18310734
|
Bio-Techne
NBP3-03700-20UL |
20 μL |
1706.22 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18335444
|
Bio-Techne
NBP3-03700-100UL |
100 μL |
3498.83 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NDUFS3 Polyclonal specifically detects NDUFS3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
NDUFS3 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
4722 | |
IgG | |
Affinity purified |
Western Blot, Immunohistochemistry, KnockDown | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
CI-30, CI-30kD, complex I 30kDa subunit, Complex I-30kD, EC 1.6.5.3, EC 1.6.99.3, EC 1.6.99.5, NADH dehydrogenase (ubiquinone) Fe-S protein 3 (30kD) (NADH-coenzyme Qreductase), NADH dehydrogenase (ubiquinone) Fe-S protein 3, 30kDa (NADH-coenzyme Qreductase), NADH dehydrogenase [ubiquinone] iron-sulfur protein 3, mitochondrial, NADH dehydrogenase-ubiquinone 30 kDa subunit, NADH-ubiquinone oxidoreductase 30 kDa subunit | |
Recombinant fusion protein containing a sequence corresponding to amino acids 37-264 of human NDUFS3 (NP_004542.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |