Learn More
Memmert™ Nearly Gastight Interior with Gas Inlet/Outlet for UF55
Nearly gastight interior with possibility for inlet/outlet through two tubes with ball valves.
Brand:  Memmert™ UF55K3
6222.44 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17569465
  • 6385.00 DKK / Each
Only null left
Estimated Shipment: 21-08-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Gas inlet/outlet through 2 tubes with ball valves
  • Technical clarification required for combination with other options
Specifications
Specifications
UF55 Oven
Interior
Gas Inlet/Outlet
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Nearly Gastight Interior with Gas Inlet/Outlet for UF55 > For Use With: UF55