Learn More
Tocris Bioscience™ Ned K
NAADP signaling inhibitor
Brand: Tocris Bioscience™ 5976/50
8779.04 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Ned K is a NAADP inhibitor. Prevents Ca2+ oscillations in rat cardiomyocytes in vitro during simulated ischemia and reperfusion, but exhibits no significant effect on Ca2+ sparks. Significantly reduces infarct size in rat cardiac ischemia/reperfusion model in vivo.Specifications
Cream | |
C31H31N5O3 | |
Soluble to 50mM in DMSO | |
Prevents Ca2+ oscillations |
Inhibitor | |
521.61g/mol | |
Store at -20°C | |
50 mg |
For Research Use Only