missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NFIL3 (Human) Recombinant Protein
Human NFIL3 full-length ORF ( AAH08197, 1 a.a. - 462 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004783-P01.25ug
This item is not returnable.
View return policy
Description
- Sequence: MQLRKMQTVKKEQASLDASSNVDKMMVLNSALTEVSEDSTTGEELLLSEGSVGKNKSSACRRKREFIPDEKKDAMYWEKRRKNNEAAKRSREKRRLNDLVLENKLIALGEENATLKAELLSLKLKFGLISSTAYAQEIQKLSNSTAVYFQDYQTSKSNVSSFVDEHEPSMVSSSCISVIKHSPQSSLSDVSEVSSVEHTQESSVQGSCRSPENKFQIIKQEPMELESYTREPRDDRGSYTASIYQNYMGNSFSGYSHSPPLLQVNRSSSNSPRTSETDDGVVGKSSDGEDEQQVPKGPIHSPVELKHVHATVVKVPEVNSSALPHKLRIKAKAMQIKVEAFDNEFEATQKLSSPIDMTSKRHFELEKHSAPSMVHSSLTPFSVQVTNIQDWSLKSEHWHQKELSGKTQNSFKTGVVEMKDSGYKVSDPENLYLKQGIANLSAEVVSLKRLIATQPISASDSG
Specifications
AAH08197 | |
nuclear factor, interleukin 3 regulated | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NFIL3 | |
GST |
4783 | |
Wheat germ expression system | |
25 μg | |
E4BP4, IL3BP1, NF-IL3A, NFIL3A | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |