missing translation for 'onlineSavingsMsg'
Learn More
Nickel(II) Nitrate Hexahydrate, purum p.a, ≥97.0% (KT), Honeywell™ Product Code.: 15616890

Nickel(II) Nitrate Hexahydrate, purum p.a, ≥97.0% (KT), Honeywell™

Product Code. 15616890 Shop All Honeywell Research Chemicals Products
1 kg, Poly bottle, 1kg
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 15616890

Brand: Honeywell Research Chemicals 722531KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purum p.a., crystallized,≥97.0%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13478-00-7
Molecular Formula H12N2NiO12
Molecular Weight (g/mol) 290.79
MDL Number MFCD00149805
InChI Key AOPCKOPZYFFEDA-UHFFFAOYSA-N
Synonym nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6
PubChem CID 61630
SMILES O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 56.7°C
Color Blue Green
pH 4.0 (at 20°C)
Physical Form Solid
Quantity 1 kg
Linear Formula Ni(NO3)2 · 6H2O
UN Number UN2725
Solubility Information 2.385g/L at 20°C
IUPAC Name nickel(2+);dinitrate;hexahydrate
Formula Weight 290.79g/mol
Percent Purity ≥97.0% (KT)
Packaging Poly bottle
Density 2.05g/mL at 25°C
Chemical Name or Material Nickel(II) nitrate hexahydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel(II) Nitrate Hexahydrate, purum p.a, ≥97.0% (KT), Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.