Learn More
Usbeck Laborgeraete™ Nickel Plated Brass Hoffman Pinchcocks
Brand: Usbeck Laborgeraete™ 5511
This item is not returnable.
View return policy
Specifications
Each | |
Hoffman Pinchcock | |
Nickel-plated Brass | |
17 mm |
Product Content Correction
Product Title
Usbeck Laborgeraete™ Nickel Plated Brass Hoffman Pinchcocks > Width: 17mm
Spot an opportunity for improvement?Share a Content Correction