missing translation for 'onlineSavingsMsg'
Learn More
(±)-Nicotine-d3 salicylate, Honeywell Fluka™ Product Code.: 15940242

(±)-Nicotine-d3 salicylate, Honeywell Fluka™

Product Code. 15940242 Shop All Honeywell Fluka Products
10 mg
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15940242

Brand: Honeywell Fluka R149510MG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Analytical standard, reference material

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY

Specifications

CAS 1173021-00-5
Packaging Glass Bottle
Quantity 10 mg
For Use With (Application) Alkaloids
Physical Form Solid
Grade Analytical Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(±)-Nicotine-d3 salicylate, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.