missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, 65% AR, Chem-Lab
Click to view available options
Quantity:
1 L
Description
The main industrial use of nitric acid is for the production of fertilizers. Nitric acid is neutralized with ammonia to give ammonium nitrate. The other main applications are for the production of explosives, nylon precursors, and specialty organic compounds.
Specifications
Specifications
pH | <1 |
Color | Colorless |
CAS | 7697-37-2 |
Melting Point | -32°C |
Boiling Point | 121°C |
Concentration | ≥65% |
Density | 1.39g/mL |
Odor | Odourless |
Vapor Pressure | 9.4hPa (20°C) |
Quantity | 1 L |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'