Learn More
Schmidlin™ NM Plus Rack Hydrogen Generator
This generator produces a continuous supply of high purity, dry hydrogen.
Brand:  Schmidlin™ DB-RNM250-EU
Encompass_Preferred
 View more versions of this product
Product Code. 17718929
  • 120107.62 DKK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Increase efficiency and improve safety and performance
  • On-demand supply 24/7
  • Pressure: 11 barg (160 psig)
  • Unique permeation membrane drying system
  • Cold dual dynamic regeneration dryer
  • USB connectivity
  • Easy to install with complete 'plug and play' functionality
  • Ideal for all GC detector applications
  • Meets and exceeds requirements for the most demanding GC applications
  • Superior hydrogen production with reliable long life cell
  • Minimal maintenance; no desiccant cartridges to change
  • PC monitoring for maintenance, diagnostics and remote connection
Specifications
Specifications
500 cm
13.3 cm
11 bar
48.5 cm
110/120 V at 60 Hz, 220/240 V at 50 Hz
125 W
250 mL/min.
35°C
19 in.
Outlet: 1/8 in.
>99.99996%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.