Learn More
NOTUM Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14663-1-AP-150UL
Description
The NOTUM antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NOTUM. This antibody recognizes human, mouse antigen. The NOTUM antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
NOTUM | |
Polyclonal | |
Unconjugated | |
150 μL | |
147111 | |
IgG |
Western Blot | |
333 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.