Learn More
Invitrogen™ NuPAGE™ Tris-Acetate SDS Buffer Kit (for Tris-Acetate Gels), Contains 1 ea. LA0041, NP0004, NP0005, NP0007
Provide separation of large molecular weight proteins when used with NuPAGE Tris-Acetate SDS Running Buffer
Brand: Invitrogen™ LA0050
1207.73 DKK valid until 2025-08-31
Use promo code "24157" to get your promotional price.
Includes: 20X NuPAGE Tris-Acetate SDS Running Buffer (500mL), 10X NuPAGE Sample Reducing Agent (250ÎĽl), NuPAGE Antioxidant (15mL), 4X NuPAGE LDS Sample Buffer (10mL)
Description
- NuPAGE Novex Tris-Acetate gels can also be run with Tris-Glycine Native Running Buffer to resolve native proteins more effectively than Tris-Glycine gel system
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Buffer Kit | |
SDS PAGE Gel | |
NuPAGE™ |
NuPAGE™ Tris-Acetate SDS Buffer Kit (for Tris-Acetate Gels) contains: • 20X NuPAGE™ Tris-Acetate SDS Running Buffer (500 ml): Store at 4°C to 25°C • 10X NuPAGE™ Sample Reducing Agent (250 μl): Store at 2°C to 8°C • NuPAGE™ Antioxidant (15 ml): Store at 4°C • 4X NuPAGE™ LDS Sample Buffer (10 ml): Store at 4°C to 25°C |
|
NuPAGE Tris-Acetate Gel | |
1 kit |
Promotions
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24157Expires: 2025-08-31