missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Tris-Glycine Mini Protein Gels, 8%, 1.0 mm, WedgeWell™ format

Invitrogen Novex Tris-Glycine Mini Gels, WedgeWell format, are polyacrylamide gels that provide reproducible separation of a wide range of proteins into well-resolved bands with two times the loading capacity.

954.00 DKK - 956.00 DKK

Specifications

For Use With (Equipment) Mini Gel Tank, XCell SureLock Mini-Cell
Gel Percentage 8%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Tris-Glycine Plus
View More Specs

Products 3
Product Code Brand Wells Price Quantity & Availability  
Product Code Brand Wells Price Quantity & Availability  
15474974
View Documents Promotion Details
Invitrogen™
XP00080BOX
10-well
954.00 DKK
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
15476794
View Documents Promotion Details
Invitrogen™
XP00082BOX
12-well
956.00 DKK
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
15486794
View Documents Promotion Details
Invitrogen™
XP00085BOX
15-well
956.00 DKK
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Invitrogen Novex Tris-Glycine Mini Gels, WedgeWell format, are polyacrylamide gels that provide reproducible separation of a wide range of proteins into well-resolved bands with two times the loading capacity. Novex Tris-Glycine Mini Gels are based on traditional Laemmli protein electrophoresis that allow the use of Laemmli sample and running buffers.

Features of Novex Tris-Glycine gels, WedgeWell format, include:
• Improved shelf life—store gels for up to 12 months at 4°C
• Diversity—use for native and denaturing protein assays
• Wedge-shaped wells—easily load up to two times more sample volume
• Fast run conditions—quickly separate your proteins using constant voltage in less than 60 minutes

Choose the right gel format for your experiments
Novex Tris-Glycine gels, WedgeWell format, come in a variety of fixed concentrations from 6% to 18%, as well as gradients with ranges of 4–12%, 4–20%, 8–16%, and 10–20%. You can select from our many well formats, including 10-, 12-, and 15-well, and different pack sizes.

Run your proteins in native or denatured form
Novex Tris-Glycine gels, WedgeWell format, do not contain SDS and can be used to run your proteins in native or in denatured form. For denatured proteins, we recommend using Novex Tris-Glycine SDS Sample Buffer (LC2676) and Novex Tris-Glycine SDS Running Buffer (LC26755). For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer (LC2673) and Novex Tris-Glycine Native Running Buffer (LC2672>.

Convenient to use
Novex Tris-Glycine gels, WedgeWell format, have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

For transferring your proteins to a membrane, we recommend using Novex Tris-Glycine Transfer Buffer (LC3675) when preforming traditional wet transfer using the Invitrogen Mini Blot Module (B1000)Transfers can also be performed with rapid semi-dry transfer using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

Specifications

Specifications

Mini Gel Tank, XCell SureLock Mini-Cell
Mini
Tris-Glycine Plus
40 to 200 kDa
Molecular Weight
10 gels (1 box)
Store at 2°C to 8°C. Do not freeze.
8 cm
8%
1.0 mm
Denaturing, Native
Denaturing, Native
Novex™, WedgeWell™
Wet Ice
8 cm
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 Months

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Novex™ Tris-Glycine Mini Protein Gels, 8%, 1.0 mm, WedgeWell™ format

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.