Learn More
Invitrogen™ ZOOM™ IPG Strips pH 9-12
Oil-free, trouble-free 2D electrophoresis in a mini-gel format
Brand:  Invitrogen™ ZM0017
Product Code. 11510916
  • 1272.00 DKK / Pack of 12
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Requires : ZOOM IPGRunner System
This item is not returnable. View return policy
Description
Description
  • First dimension separation, isoelectric focusing (IEF), is complete in less than three hours
  • The mini-gel design of the system is easy to handle, eliminates mineral oil overlays, and allows processing of up to 12 samples at once for high-throughput usage
  • Strips are 7cm long and contain a thin layer of polyacrylamide gel that includes a fixed pH gradient
  • Each strip is clearly labeled with a unique identifying number, pH range, and orientation marks
  • Strips are supplied attached to a tri-fold card for easy access and removal
2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Dry Ice
TRUSTED_SUSTAINABILITY
Specifications
Specifications
The ZOOM™ IPGRunner™ Mini-Cell includes the ZOOM™ IPGRunner™ Buffer Core and Lid, Gel Tension Wedge, Buffer Dam, and the Mini- Cell chamber. The ZOOM™ IPGRunner™ Combo Kit includes the ZOOM™ IPGRunner™ Mini-Cell, electrode wicks, sealing tape, 10 ZOOM™ IPGRunner™ Cassettes, and 12 ZOOM™ Strips, pH 3-10 NL. The ZOOM™ IPGRunner™ Retrofit Kit for XCell SureLock™ Mini-Cell includes the ZOOM™ IPGRunner™ Buffer Core and Lid. The ZOOM™ IPGRunner™ Cassettes include electrode wicks and sealing tape.
Mini
IPG Strip
12 strips
XCell SureLock Mini-Cell
9.0 - 12.0
ZOOM™
Dry Ice
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
For Research Use Only. Not for use in diagnostic procedures.