missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR2E3 Mouse anti-Human, Clone: 1G1D9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66494-1-IG-20UL
This item is not returnable.
View return policy
Description
NR2E3, a NR2 Hepatocyte NF4-Like Receptor, has been suggested to affect cone cell proliferation in eye development. It has been shown to have a role in determining photoreceptor phenotype during human retinogenesis. In RD7 mice, a deletion in NR2E3 results in hereditary retinal dysplasia and degeneration. A homozygous missense mutation in NR2E3 has been linked to retinitis pigmentosa in Crypto-Jews of Belmonte. Two alternatively spliced isoforms have been documented. NR2E3 is expressed in the retina. ESTs have been isolated from cancerous human lung, colon, and ovary, and from normal human eye.Specifications
NR2E3 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y5X4 | |
NR2E3 | |
NR2E3 Fusion Protein Ag5831 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
1G1D9 | |
Unconjugated | |
NR2E3 | |
ESCS, NR2E3, PNR, rd7, RNR, RP37 | |
Mouse | |
Protein G | |
RUO | |
10002 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NR2E3 Mouse anti-Human, Clone: 1G1D9, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction