missing translation for 'onlineSavingsMsg'
Learn More
NS 3623, Tocris Bioscience™ Product Code.: 18094542

NS 3623, Tocris Bioscience™

Product Code. 18094542
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18094542

Brand: Tocris Bioscience 4462/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KV11.1 (hERG) and KV4.3 channel activator; antiarrhythmic

KV11.1 (hERG) and KV4.3 channel activator. Activates the IKr and Ito currents and displays antiarrhythmic activity. May also act as a partial blocker of KV11.1 channels. Displays selectivity over the key cardiac potassium channels, KV7.1 (KCNQ1) and KV1.5. Suitable for both in vitro and in vivo use.

Chemical Identifiers

CAS 343630-41-1
Molecular Formula C15H10BrF3N6O
Molecular Weight (g/mol) 427.185
InChI Key JXPULDIATMTIIN-UHFFFAOYSA-N
PubChem CID 9954236
IUPAC Name 1-[4-bromo-2-(2H-tetrazol-5-yl)phenyl]-3-[3-(trifluoromethyl)phenyl]urea
SMILES C1=CC(=CC(=C1)NC(=O)NC2=C(C=C(C=C2)Br)C3=NNN=N3)C(F)(F)F

Specifications

Quantity 10 mg
Synonym d0jy2k, 1-4-bromo-2-2h-tetrazol-5-yl phenyl-3-3-trifluoromethyl phenyl urea, i kr agonists arrhythmia , neurosearch, herg potassium channel openers arrhythmia, herg potassium channel openers arrhythmia, n-4-bromo-2-1h-tetrazol-5-yl-phenyl-n'-3'-trifluoromethylphenyl urea, n-4-bromo-2-1h-tetrazol-5-yl-phenyl-n'-3-trifluoromethyl phenyl-urea, rapid delayed-rectifier current activators acquired long qt syndrome, 1-4-bromo-2-1h-1,2,3,4-tetrazol-5-yl phenyl-3-3-trifluoromethyl phenyl urea
Formula Weight 427.18
Percent Purity >99%
Chemical Name or Material NS 3623
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
NS 3623, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.