Beskrivelse
NSC 23766 is a selective inhibitor of Rac1-GEF interaction. NSC 23766 prevents Rac1 activation by Rac-specific guanine nucleotide exchange factors (GEFs) TrioN and Tiam1 (IC50 ∼ 50 μM) without affecting Cdc42 or RhoA activation. The compound inhibits Rac1-mediated cell functions and is reported to reverse tumor cell phenotypes in prostate cancer cells. NSC 23766 also inhibits mammary carcinoma cell invasion in vitro and inhibits mammary tumor progression and metastasis in a mouse model.
Greener Choice-anprisninger
- undefined
- Ozone-Safe or Safe for the Ozone Layer
Tekniske data
Tekniske data
Mængde | 50 mg |
Produkttype | NSC 23766 |
Renhed | >98% |
Molekylvægt (g/mol) | 530.97 |
Inhibitorer | Rho GTPase Inhibitors |
For Research Use Only
Korrektion af produktindhold
Produkttitel
Tocris Bioscience™ NSC 23766 >
Ser du en mulighed for forbedring?Del en indholdskorrektion