Learn More
Nuclear Receptor Buffer F
Nuclear Receptor Buffer F
Brand:  Invitrogen PV4547
Product Code. 10116403
  • 1104.00 DKK / Each
Explore more special offers
Description
Description
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nuclear Receptor Buffer
25 mL
Liquid
Nuclear Receptor Buffer F may be shipped frozen or at room temperature. It should be stored at room temperature (20-30°C).
Approved for shipment at Room Temperature or on Wet or Dry Ice
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2934123B Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberPV4547
Lot Number2970822C Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberPV4547
Lot Number2970822B Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberPV4547
Lot Number2970822A Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberPV4547
Lot Number2934123A Certificate TypeCertificate of Analysis Date25/04/2024 Catalog NumberPV4547
Product Title
Nuclear Receptor Buffer F > 25mL