Learn More
Thermo Scientific™ 384 Well White Plate, TC Surface, Pack of 10
Color:White; Sterility:Sterile; Lid:With Lid
Brand: Thermo Scientific™ 164610
2823.47 DKK valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
Improve background, autofluorescence and crosstalk with Thermo Scientific™ Nunc 384-Well Polystyrene Black and White Microplates.
- Black microplates are recommended for fluorescence measurements; with minimum back-scattered light and background fluorescence
- White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
0.084 cm2/well | |
10 Pk., 30 Cases | |
White | |
Polystyrene | |
384 | |
Sterile | |
Flat Bottom (F-well) | |
10 |
With Lid | |
16 x 24 | |
Nunc | |
Microplate | |
120 ÎĽL | |
10 to 100 ÎĽL | |
Cell Culture | |
30 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1420897 | Certificate of Processing | 21/04/2025 | 164610 |
1420895 | Certificate of Processing | 13/02/2025 | 164610 |
1420896 | Certificate of Processing | 12/02/2025 | 164610 |
1425083 | Certificate of Compliance | 02/04/2025 | 164610 |
1425082 | Certificate of Compliance | 01/04/2025 | 164610 |