Learn More
Thermo Scientific™ Cryo Vial Closure Color Coders
GREENER_CHOICE
Make it easier to identify specific vials quickly and simplify your inventory by using these color coders that fit into the top of cryogenic vial or tube closures.
Brand:  Thermo Scientific™ 5045-0003
74.19 DKK valid until 2025-12-31
Use promo code "24325" to get your promotional price.
 View more versions of this product
Product Code. 10450423
  • 128.00 DKK / Pack of 100
Cryo Collection Promotion
Use promo code 24082 at checkout
View Available Promotions
Only null left
In Stock
Description
Make it possible to identify specific vials quickly and simplify your inventory using color coders that fit into the top of Nalgene™ Cryogenic vial closures or Nunc™ Cryogenic Tube closures.
  • Nalgene™ color coders are made from polystyrene and fit only in Nalgene™ cryogenic vial closures. Available in 5 colors
  • Nunc™ color coders are made from polycarbonate and fit only in Nunc™ cryogenic tube closures and Cryobank 2ml and 5mL tube caps. Available in 10 colors or as an assortment.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nalgene™ Color coder
Polycarbonate
100/Pk., 500/Cs.
Blue
Nalgene
Vial Closure
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1431063 Certificate TypeCertificate of Compliance Date02/07/2025 Catalog Number5045-0003
Lot Number1422981 Certificate TypeCertificate of Compliance Date19/02/2025 Catalog Number5045-0003
Lot Number1417358 Certificate TypeCertificate of Compliance Date12/11/2024 Catalog Number5045-0003
Lot Number1414549 Certificate TypeCertificate of Compliance Date21/09/2024 Catalog Number5045-0003
Lot Number1407796 Certificate TypeCertificate of Compliance Date05/06/2024 Catalog Number5045-0003
Promotions
Promotions
  • Thermo Scientific Nunc , Thermo Scientific Nalgene , Thermo Scientific Abgene Stora
    Save up to 60% on cold storage essentials.
    Promo Code: 24082
    Expires2025-12-31
    Days
    160
    Hours
    23
    Minutes
    23
    Seconds
    3
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24157
    Expires2025-08-31
    Days
    38
    Hours
    23
    Minutes
    23
    Seconds
    3
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/07/2025 and 31/08/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Thermo Scientific™ Cryo Vial Closure Color Coders > Nalgene Cryovial color coder, PS, blue