Learn More
Cytiva (formerly Pall Lab) Nylaflo™ Nylon Membrane Disc Filters
A highly versatile laboratory membrane filter.
1050.00 DKK - 5095.00 DKK
Product Code | Brand | Pore Size | Diameter (Metric) Membrane | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Diameter (Metric) Membrane | Price | Quantity & Availability | ||||
17144591
|
Cytiva (formerly Pall Lab)
66609 |
0.45 μm | 90 mm |
5095.00 DKK
Pack of 100 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17184581
|
Cytiva (formerly Pall Lab)
66602 |
0.2 μm | 47 mm |
2055.00 DKK
Pack of 100 |
Estimated Shipment: 02-01-2025
Log in to see stock. |
||||
17104591
|
Cytiva (formerly Pall Lab)
66604 |
0.2 μm | 142 mm |
2790.00 DKK
Pack of 25 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
17114591
|
Cytiva (formerly Pall Lab)
66606 |
0.45 μm | 13 mm |
1050.00 DKK
Pack of 100 |
Estimated Shipment: 28-11-2024
Log in to see stock. |
||||
17124591
|
Cytiva (formerly Pall Lab)
66607 |
0.45 μm | 25 mm |
1185.00 DKK
Pack of 100 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
17134591
|
Cytiva (formerly Pall Lab)
66608 |
0.45 μm | 47 mm |
1920.00 DKK
Pack of 100 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||||
17174581
|
Cytiva (formerly Pall Lab)
66601 |
0.2 μm | 25 mm |
1235.00 DKK
Pack of 100 |
Estimated Shipment: 28-11-2024
Log in to see stock. |
||||
17154591
|
Cytiva (formerly Pall Lab)
66610 |
0.45 μm | 142 mm |
3080.00 DKK
Pack of 25 |
Estimated Shipment: 25-11-2024
Log in to see stock. |
||||
17194581
|
Cytiva (formerly Pall Lab)
66603 |
0.2 μm | 90 mm |
5095.00 DKK
Pack of 100 |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||||
Description
- Chemical compatibility with esters, bases, and alcohols naturally hydrophilic
- HPLC certified
- Provides assurance that the filter will not add artifacts to your analysis
- For research use only
Specifications
Nylon (Hydrophilic) | |
Non-sterile |