missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ O-GlcNAcase/OGA/MGEA5 293T Cell Transient Overexpression Lysate
Overexpression Lysate
Brand: Novus Biologicals™ H00010724-T01
This item is not returnable.
View return policy
Specifications
0.1 mL | |
O-GlcNAcase/OGA/MGEA5 | |
OGA |
Store at -80°C. Avoid freeze-thaw cycles. | |
bifunctional protein NCOAT, FLJ11229, HEXC, hyaluronidase in meningioma, KIAA0679, MEA5FLJ23355, meningioma expressed antigen 5 (hyaluronidase), Meningioma-expressed antigen 5, NCOAT, Nuclear cytoplasmic O-GlcNAcase and acetyltransferase, OGA, O-GlcNAcase |