Learn More
DWK Life Sciences O-ring, made of nitrile, for safety standard ground sockets
Component of the DURAN™ safety ground joint.
44.90 DKK - 121.00 DKK
Specifications
Material | Nitrile |
---|---|
For Use With (Application) | To secure the security grindings; Sealing |
Quantity | 10/Pk. |
Product Code | Brand | Joint Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | ||||||
15921561
|
DWK Life Sciences
286217705 |
NS 45/40 |
121.00 DKK
Pack of 10 |
Estimated Shipment: 03-02-2025
Log in to see stock. |
||||||
15864608
|
DWK Life Sciences
286216803 |
NS 14/23 |
44.90 DKK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15991551
|
DWK Life Sciences
286217002 |
NS 19/26 |
63.25 DKK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15884608
|
DWK Life Sciences
286217405 |
NS 29/32 |
89.65 DKK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15894608
|
DWK Life Sciences
286217508 |
NS 34/35 |
96.15 DKK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15911561
|
DWK Life Sciences
286217208 |
NS 24/29 |
87.00 DKK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
Description
Nitrile O-rings for DURAN™ safety ground joints together with the screw cap secure the connection between core and sleeve.
- In combination with the screw cap, the safety ground joints (core and sleeve) can be sealed tightly.
Specifications
Nitrile | |
To secure the security grindings; Sealing | |
10/Pk. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
DWK Life Sciences O-ring, made of nitrile, for safety standard ground sockets