missing translation for 'onlineSavingsMsg'
Learn More

Octane, Reagent Grade, 98%, Honeywell™

Product Code. 15615450
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-65-9
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00009556
InChI Key TVMXDCGIABBOFY-UHFFFAOYSA-N
Synonym n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
PubChem CID 356
ChEBI CHEBI:17590
IUPAC Name octane
SMILES CCCCCCCC

Specifications

Melting Point -57°C
Density 0.703g/mL at 25°C
Boiling Point 125°C to 127°C
Flash Point 13°C (55.4°F)
Infrared Spectrum Conforms to structure
Packaging Glass Bottle
Linear Formula CH3(CH2)6CH3
Refractive Index n20/D 1.398
Quantity 100 mL
UN Number UN1262
Beilstein 1696875
Vapor Density 3.9 (vs air)
Formula Weight 114.23g/mol
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Octane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.