Learn More
OHAUS™ Triple-Beam Balances:Polypropylene Scoop for Balances without Tare
Scoop with counterweight is footed to prevent spills
Brand: OHAUS™ 80780014
This item is not returnable.
View return policy
Description
For holding and pouring granular materials. Scoop with counterweight is footed to prevent spills.
Order Info
L x W x H: 12 x 6 x 2 3/4 in. (30 x 15 x 70cm).
Specifications
Scoop | |
Scoop and Counter Weight, Poly | |
Footed | |
Holding and storing granular materials | |
12 x 6 x 2.75 in. (30.5 x 15.24 x 6.98 cm) | |
6.98 cm | |
30.5 cm | |
15.24 cm |
Polypropylene | |
24g counterweight | |
White | |
Ohaus™ triple-beam dial-O-gram 2000 balances | |
2.75 in. | |
12 in. | |
6 in. |
Product Content Correction
Product Title
OHAUS™ Triple-Beam Balances:Polypropylene Scoop for Balances without Tare > 4 x 3.5 in. (10.2 cm X 8.9 cm); Weight: 24g
Spot an opportunity for improvement?Share a Content Correction