Learn More
OHAUS™ USB Cable for Explorer Balances
Allows Explorer balances to be connected to peripherals, including monitors and computers.
Brand:  OHAUS™ 83021085
174.59 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11840133
  • 180.00 DKK / Each
Explore more special offers
Description
Description
USB Cable (Type A to B)
Specifications
Specifications
Explorer Balance
Videos
  • Ohaus - Explorer balances
    Ohaus - Explorer balances
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Ohaus Explorer balances
    Product Title
    OHAUS™ USB Cable for Explorer Balances > USB Interface cable for Explorer balance