missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ OLIG1 HEK293 Cell Transient Overexpression Lysate (Nondenatured)
Human overexpression lysate
Brand: Abnova™ L002T6.100ug
This item is not returnable.
View return policy
Specifications
HEK293 | |
OLIG1 HEK293 Cell Transient Overexpression Lysate(Non-Denatured) | |
Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. | |
pCMV-OLIG1 full length | |
100 μg | |
Human | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunoprecipitation, Western Blot | |
NM_138983.1 | |
Theoretical M.W. 27.905kDa | |
Transient overexpression cell lysate was tested with Anti-OLIG1 antibody by Western Blots. | |
In modified RIPA Lysis Buffer. | |
2 mg/mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 100UG OLIG1 HEK293 Cell Transient Overexpression L
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Mercaptoethanol