missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jaytec Glass™ Open Top Borosilicate Glass Dropping Funnels
Open Top Borosilicate Glass Dropping Funnels for easy refilling in situ. The stopcock permits carefully controlled flow of reagents.
Brand: Jaytec Glass™ FPM-210-030D
This item is not returnable.
View return policy
Description
- Open top
- Cylindrical shape
- With interchangeable PTFE stopcock
Specifications
24 mm | |
8 mm | |
232 mm | |
PTFE stopcock key | |
Dropping funnel |
25 mL | |
Borosilicate Glass | |
No | |
Yes |