Description
- For opus™
- Connection cable between module and base unit
Specifications
Specifications
For Use With (Application) | Starting the Dosing Operation Externally (for Basic Units 10 + 20 + 50 mL), e.g. For the Integration in Automatic Processes |
No. per Pack | 1 Each |
Type | Control Cable |
Product Content Correction
Product Title
Hirschmann™ Operating Cable >
Spot an opportunity for improvement?Share a Content Correction