Learn More
Hirschmann Instruments™ opus™ Dispenser
Guarantees excellent accuracy during serial dispensing. Hirschmann Instruments™ opus™ Dispenser is for motor-driven dispensing directly from the reagent bottle.
Brand: Hirschmann Instruments™ 9581020
14465.77 DKK valid until 2025-07-31
Use promo code "24075" to get your promotional price.
Description
- Dispenser touch screen module incl. brown UV protection
- With thread A 45mm, 3 adapters, 1 dispensing discharge unit, 1 power supply
- With reagent recirculation system
- Conformity certified
- Serial and USB interface
Specifications
20 mL | |
TFT touchscreen | |
100/240 V |
Promotions
-
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-07-31