Description
- Provides for hands-free operation
- Variable switch programming
- Accessory item
- Suitable for Opus™ dispensers and titration units
- Connecting cable included
Specifications
Specifications
Color | Black, Blue |
For Use With (Equipment) | Opus Dispensers and Titration Units |
For Use With (Application) | Hands-Free Operation |
Includes | Connecting Cable |
Type | Foot Key |
Product Content Correction
Product Title
Hirschmann™ Opus™ Pedal Switch >
Spot an opportunity for improvement?Share a Content Correction