missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORAI1 Mouse anti-Human, Clone: 3C12F9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66223-1-IG-20UL
This item is not returnable.
View return policy
Description
ORAI1 is a plasma membrane protein essential for store-operated calcium entry.Specifications
ORAI1 | |
Monoclonal | |
1.02 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96D31 | |
Mouse | |
Protein A | |
RUO | |
84876 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
3C12F9 | |
Unconjugated | |
ORAI1 | |
ORAI1 | |
ORAI1 Fusion Protein Ag18050 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ORAI1 Mouse anti-Human, Clone: 3C12F9, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction