Learn More
Bochem™ Orange Plastic Stool
Click to view available options
Color:
Orange
Diameter (Metric) Base:
450 mm
Height (Metric):
420 mm
Material:
Plastic
Unit Size:
Each
Description
- GS safety control
Specifications
Specifications
Product Type | Kick Stool |
Color | Orange |
Diameter (Metric) Base | 450 mm |
Diameter (Metric) | 300 mm |
Height (Metric) | 420 mm |
Quantity | 1 Ea. |
Material | Plastic |
Product Content Correction
Product Title
Bochem™ Orange Plastic Stool >
Spot an opportunity for improvement?Share a Content Correction