Learn More
Honeywell Fluka™ 6-Chloro-2,4-diamino-1,3,5-triazine-13C3
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Specifications
Specifications
Quantity | 10 mg |
Packaging | Glass Bottle |
For Use With (Application) | TRIAZINES |
CAS | 1216850-33-7 |
Product Content Correction
Product Title
Honeywell Fluka™ 6-Chloro-2,4-diamino-1,3,5-triazine-13C3 >
Spot an opportunity for improvement?Share a Content Correction